Thermo Scientific Chemicals

3-Chlorocinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog number: A13416.09
10 g, Each
Thermo Scientific Chemicals

3-Chlorocinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog number: A13416.09
10 g, Each
Quantity
Catalog number: A13416.09
also known as A13416-09
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
1866-38-2
IUPAC Name
(2E)-3-(3-chlorophenyl)prop-2-enoic acid
Molecular Formula
C9H7ClO2
InChI Key
FFKGOJWPSXRALK-SNAWJCMRSA-N
SMILES
OC(=O)C=CC1=CC=CC(Cl)=C1
Form
Crystals or powder or crystalline powder or fused solid
Assay (HPLC)
≥98.0%
Melting Point (clear melt)
159.0-166.0?C
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text