2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%
Thermo Scientific Chemicals

2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%

CAS: 7128-64-5 | C26H26N2O2S | 430.566 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14928.14
also known as A14928-14
Price (EUR)
102,65
Online Exclusive
114,00
Save 11,35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
102,65
Online Exclusive
114,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS7128-64-5
IUPAC Name5-tert-butyl-2-[5-(5-tert-butyl-1,3-benzoxazol-2-yl)thiophen-2-yl]-1,3-benzoxazole
Molecular FormulaC26H26N2O2S
InChI KeyAIXZBGVLNVRQSS-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C2OC(=NC2=C1)C1=CC=C(S1)C1=NC2=CC(=CC=C2O1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)199.0-205.0?C
Identification (FTIR)Conforms
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene is used as an optical brightener which converts UV light into visible light. It is also used as a fluorescent brightener for thermoplastic resins of polyvinyl chloride, acrylic resin, polyester fiber, paint, coating and printing ink. It is also useful in detecting the presence of thin-film electrical contact lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene is used as an optical brightener which converts UV light into visible light. It is also used as a fluorescent brightener for thermoplastic resins of polyvinyl chloride, acrylic resin, polyester fiber, paint, coating and printing ink. It is also useful in detecting the presence of thin-film electrical contact lubricants.

Solubility
Soluble in acetone and toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Fourati, M. A.; Maris, T.; Skene, W. G.; Bazuin, C G.; Prud’homme, R. E. Photophysical, Electrochemical and Crystallographic Investigations of the Fluorophore 2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene. J. Phys. Chem. B 2011, 115 (43), 12362-12369.
  2. Giani, M.; Sist, P.; Berto, D.; Serrazanetti, G. P.; Ventrella, V.; Urbani, R. The organic matrix of pelagic mucilaginous aggregates in the Tyrrhenian Sea (Mediterranean Sea). Mar. Chem. 2012, 132, 83-94.