2-Ethoxybenzaldehyde, 97+%
2-Ethoxybenzaldehyde, 97+%
2-Ethoxybenzaldehyde, 97+%
Thermo Scientific Chemicals

2-Ethoxybenzaldehyde, 97+%

CAS: 613-69-4 | C9H10O2 | 150.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
Catalog number A15035.22
also known as A15035-22
Price (EUR)
75,60
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
75,60
Each
Chemical Identifiers
CAS613-69-4
IUPAC Name2-ethoxybenzaldehyde
Molecular FormulaC9H10O2
InChI KeyDUVJMSPTZMCSTQ-UHFFFAOYSA-N
SMILESCCOC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤2.0%
Appearance (Color)Clear colorless to yellow or yellow-brown
Assay (GC)≥97.0%
Refractive Index1.5405-1.5455 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only