Bromotrimethylsilane, 97%, stab. with copper powder or silver wire
Bromotrimethylsilane, 97%, stab. with copper powder or silver wire
Bromotrimethylsilane, 97%, stab. with copper powder or silver wire
Thermo Scientific Chemicals

Bromotrimethylsilane, 97%, stab. with copper powder or silver wire

CAS: 2857-97-8 | C3H9BrSi | 153.094 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A15334.09
also known as A15334-09
Price (EUR)
42,40
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
42,40
Each
Chemical Identifiers
CAS2857-97-8
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
Refractive Index1.4220-1.4280 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.

Solubility
Miscible with carbon tetrachloride, chloroform, dichloroethane, acetonitrile, toluene, dichloromethane and hexane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids, water and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Silylating agent; see Appendix 4. About 104x more reactive than TMSCl in the conversion of ketones to their silyl enol ethers in the presence of Et3N: Liebigs Ann. Chem., 1718 (1980).
  2. Converts alcohols to alkyl bromides: Tetrahedron Lett., 4483 (1978), and acyl chlorides to acyl bromides: Synthesis, 216 (1981).
  3. Acetals, including MOM and THP ethers are readily cleaved, as are trityl and TBDMS ethers: Tetrahedron Lett., 25, 2515 (1984). Methyl glycosides are converted to the glycosyl bromides: Tetrahedron Lett., 22, 513 (1981).
  4. Epoxides, oxetanes and THF are cleaved by the reagent: Synthesis, 383 (1981), as are small ring lactones: Angew. Chem. Int. Ed., 18, 689 (1979). The cleavage of acyclic ethers and esters is markedly catalyzed by IBr: J. Org. Chem., 48, 1678 (1983).
  5. In combination with DMSO and Et3N, effects the bromolactonization of unsaturated acids: J. Chem. Soc., Perkin 1, 847 (1994).
  6. Effective catalyst for Michaelis-Arbuzov rearrangement of trivalent organophosphorus P-O-R compounds to the corresponding P=O derivatives: Org. Lett., 5, 1661 (2003).
  7. Compare also Iodotrimethyl silane, A12902.
  8. Ferry, L.; Dorez, G.; Taguet, A.; Otazaghine, B.; Lopez-Cuesta, J. M. Chemical modification of lignin by phosphorus molecules to improve the fire behavior of polybutylene succinate. Polym. Degrad. Stab. 2015, 113, 135-143.
  9. Willkomm, J.; Muresan, N. M.; Reisner, E. Enhancing H2 evolution performance of an immobilised cobalt catalyst by rational ligand design. Chem. Sci. 2015, 6, 2727-2736.