Copper(II) oxalate hemihydrate, 98%
Copper(II) oxalate hemihydrate, 98%
Copper(II) oxalate hemihydrate, 98%
Thermo Scientific Chemicals

Copper(II) oxalate hemihydrate, 98%

CAS: 5893-66-3 | C4H8Cu2O10 | 343.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A15365.30
also known as A15365-30
Price (EUR)
128,65
線上優惠
143,00
Save 14,35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
128,65
線上優惠
143,00
Save 14,35 (10%)
Each
Chemical Identifiers
CAS5893-66-3
SpecificationsSpecification SheetSpecification Sheet
Trace MetalCu: ≥38.5% (theory 39.58%)
Trace MetalNa: < 0.5%
Appearance (Color)Pale blue
Assay (unspecified)>97.5% by elemental copper
Trace MetalFe: < 100ppm
View more
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

Solubility
Soluble in NH{4}OH

Notes
Store in cool, dry, and well-ventilated area. Store away from oxidizing agents. Incompatible with acitve metals.
RUO – Research Use Only

General References:

  1. Jianfang Cao; Shuxia Liu; Ruige Cao; Linhua Xie; Yuanhang Ren; Chaoying Gao and Lin Xu. Organic-inorganic hybrids assembled by bis(undecatungstophosphate) lanthanates and dinuclear copper(II)-oxalate complexes.Dalton Trans.2008, 115-120.
  2. Kenneth T. McGregor; Zoltan G. Soos. Anisotropic exchange and temperature-dependent electron paramagnetic resonance line width in one-dimensional copper(II) complexes. 2. Magnetic properties of copper(II) oxalate-1/3-water.Inorg. Chem.1976, 15 (9), 2159-2165.