4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%
4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%
4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%
4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%

CAS: 94108-56-2 | C7H4ClF3O3S | 260.611 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15467.06
also known as A15467-06
Price (EUR)
82,60
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
82,60
Each
Add to cart
Chemical Identifiers
CAS94108-56-2
IUPAC Name4-(trifluoromethoxy)benzene-1-sulfonyl chloride
Molecular FormulaC7H4ClF3O3S
InChI KeyUHCDBMIOLNKDHG-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4775-1.4815 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only