3-Methoxybenzyl alcohol, 97+%
3-Methoxybenzyl alcohol, 97+%
3-Methoxybenzyl alcohol, 97+%
3-Methoxybenzyl alcohol, 97+%
Thermo Scientific Chemicals

3-Methoxybenzyl alcohol, 97+%

CAS: 6971-51-3 | C8H10O2 | 138.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A15471.22
also known as A15471-22
Price (EUR)
93,50
Online Exclusive
110,00
Save 16,50 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
93,50
Online Exclusive
110,00
Save 16,50 (15%)
Each
Add to cart
Chemical Identifiers
CAS6971-51-3
IUPAC Name(3-methoxyphenyl)methanol
Molecular FormulaC8H10O2
InChI KeyIIGNZLVHOZEOPV-UHFFFAOYSA-N
SMILESCOC1=CC=CC(CO)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow to pale brown
FormLiquid
Assay (GC)≥97.0%
3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in ether, and alcohol. Insoluble in water.

Notes
Incompatible with Oxidizing agents, acids, acid chlorides, acid anhydrides.
RUO – Research Use Only
Nicholas J. Turro; Peter Wan. Fluorescence quenching of methoxybenzyl alcohols by the hydronium ion in aqueous solution: acid-catalyzed formation of methoxybenzyl cations from the singlet excited state.Journal of photochemistry.1985, 28 93-102.