Sulfur, flowers, 99%
Sulfur, flowers, 99%
Sulfur, flowers, 99%
Thermo Scientific Chemicals

Sulfur, flowers, 99%

CAS: 7704-34-9 | S
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A17260.36
also known as A17260-36
Price (EUR)
39,50
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
39,50
Each
Chemical Identifiers
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (unspecified)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.