3-Chloro-4-hydroxybenzaldehyde, 98%
3-Chloro-4-hydroxybenzaldehyde, 98%
3-Chloro-4-hydroxybenzaldehyde, 98%
Thermo Scientific Chemicals

3-Chloro-4-hydroxybenzaldehyde, 98%

CAS: 2420-16-8 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17515.14
also known as A17515-14
Price (EUR)
330,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
330,00
Each
Chemical Identifiers
CAS2420-16-8
IUPAC Name3-chloro-4-hydroxybenzaldehyde
Molecular FormulaC7H5ClO2
InChI KeyVGSOCYWCRMXQAB-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)128.0-134.0?C
Appearance (Color)Pale cream to cream or pale yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only