Thermo Scientific Chemicals

Murexide, Thermo Scientific Chemicals

Catalog number: A17540.18
50 g, Each
Thermo Scientific Chemicals

Murexide, Thermo Scientific Chemicals

Catalog number: A17540.18
50 g, Each
Quantity
Catalog number: A17540.18
also known as A17540-18
Price (EUR)
Price: 151,00
Your price:
Quantity
-

Chemical Identifiers

CAS
3051-09-0
IUPAC Name
ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide
Molecular Formula
C8H8N6O6
InChI Key
PCCZWUCAYILEMX-UHFFFAOYSA-O
SMILES
[NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1
Form
powder
Appearance (Color)
Dark red or purple
lambda-max (UV-Vis)
520 - 525nm (water)
Comment
Suitable for complexometric titrations

Description

Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.

Solubility
Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.                  
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text