Hexafluoroglutaric acid, 98%
Hexafluoroglutaric acid, 98%
Hexafluoroglutaric acid, 98%
Hexafluoroglutaric acid, 98%
Thermo Scientific Chemicals

Hexafluoroglutaric acid, 98%

CAS: 376-73-8 | C5H2F6O4 | 240.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17848.06
also known as A17848-06
Price (EUR)
52,20
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
52,20
Each
Add to cart
Chemical Identifiers
CAS376-73-8
IUPAC Namehexafluoropentanedioic acid
Molecular FormulaC5H2F6O4
InChI KeyCCUWGJDGLACFQT-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only