L-Tyrosine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
L-Tyrosine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
L-Tyrosine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Tyrosine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A18358.14
also known as A18358-14
25g
A18358.22
also known as A18358-22
100g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A18358.14
also known as A18358-14
Price (EUR)
57,30
Each
Quantity:
25g
Request bulk or custom format
Price (EUR)
57,30
Each
Chemical Identifiers
CAS4089-07-0
IUPAC Nameethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
Molecular FormulaC11H16ClNO3
InChI KeyBQULAXAVRFIAHN-OXZMBGLCNA-N
SMILESCl.CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-5.5° to -8.5 ° (c=2 in water)
Appearance (Color)White to cream
FormPowder or crystalline powder or crystals
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Melting Point (clear melt)164.0-174.0°C
It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.

Solubility
Soluble in water (3.685e+005 mg/L @ 25°C (est.)).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Tito A.; Jimenez-Lopez C, et al. A study of the intermolecular interactions of tolmetin-acetyl-L-tyrosine ethyl ester complex.Spectrochim. Acta A.2009,721000-1006.

Feda E Ali.; Andrew Leung.; Robert A Cherny.; Christine Mavros.; Kevin J Barnham.; Frances Separovic.; Colin J Barrow. Dimerisation of N-acetyl-L-tyrosine ethyl ester and Abeta peptides via formation of dityrosine.Free Radic Res.2006,40(1), 1-9.