Perfluoro(2-methyl-2-pentene), 97%
Perfluoro(2-methyl-2-pentene), 97%
Perfluoro(2-methyl-2-pentene), 97%
Perfluoro(2-methyl-2-pentene), 97%
Thermo Scientific Chemicals

Perfluoro(2-methyl-2-pentene), 97%

CAS: 1584-03-8 | C6F12 | 300.047 g/mol
Have Questions?
Catalog NumberQuantity
A18394.14
also known as A18394-14
25 g
Catalog number A18394.14
also known as A18394-14
Price (EUR)
302,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
302,00
Each
Add to cart
Chemical Identifiers
CAS1584-03-8
IUPAC Name1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene
Molecular FormulaC6F12
InChI KeyFAEGGADNHFKDQX-UHFFFAOYSA-N
SMILESFC(=C(C(F)(F)F)C(F)(F)F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Indexca 1.3000 @ 20?C
Appearance (Color)Clear, colorless
CommentMain contaminant Perfluoro(4-methyl-2-pentene), ca 2-3%
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only