L-Threonine
Ajinomoto Group

L-Threonine

L-threonine is an essential amino acid that serves critical metabolic and structural functions in biological systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
A2131450 kgFiber Drum
A346011 kgPoly Drum
A3460210 kgPoly Drum
Catalog number A21314
Price (EUR)
-
Quantity:
50 kg
Packaging:
Fiber Drum

L-threonine is an essential amino acid that serves critical metabolic and structural functions in biological systems. As a precursor for the biosynthesis of glycine and serine, threonine plays a fundamental role in amino acid metabolism. The compound functions as a key structural component in collagen formation and enamel protein synthesis, while simultaneously facilitating hepatic fat metabolism. Additionally, L-threonine enhances nutrient bioavailability by improving absorption efficiency when present in formulations.

This pharmaceutical-grade L-threonine is manufactured by Ajinomoto Health & Nutrition North America, Inc., leveraging advanced biotechnology processes to deliver superior product quality for biotech/pharmaceutical applications, cell culture media formulations, and intravenous therapeutic preparations.

Complies with United States Pharmacopeia (USP), European Pharmacopoeia (EP), and Japanese Pharmacopoeia (JP) standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS72-19-5
Chemical Name or MaterialL-Threonine
PackagingFiber Drum
GradeEP/JP/USP
Molecular Weight (g/mol)119.12
Percent Purity99.0 to 101.0%
Quantity50 kg
Unit SizeEach