B-27™ Supplement (50X), gamma irradiated, minus insulin
B-27™ Supplement (50X), gamma irradiated, minus insulin
Gibco™
B-27™ Supplement (50X), gamma irradiated, minus insulin
B-27 Supplement minus insulin is a serum-free supplement designed to support growth of hippocampal and other CNS neurons in applicationsRead more
Have Questions?
Catalog NumberQuantity
A46746SA10 mL
Catalog number A46746SA
Price (EUR)
-
Quantity:
10 mL
B-27 Supplement minus insulin is a serum-free supplement designed to support growth of hippocampal and other CNS neurons in applications where the presence of insulin causes interference.
Gamma irradiated B-27 Supplement minus insulin has the same formulation as standard B-27 Supplement minus insulin. After the product is manufactured, an irradiation process is conducted at a facility that complies with the ISO 11137 standard. In a study verified by the PMDA, it was confirmed that this irradiation process (of 25 kGy or more) does inactivate/remove viruses.
Please contact your local sales representatives for seeking the Certificate of Irradiation.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeNeural Cells
Concentration50 X
Culture TypeMammalian Cell Culture
Product LineB-27
Quantity10 mL
Serum LevelSerum-Free
Shipping ConditionDry Ice
Tested ForMycoplasma, Endotoxin, Sterility
ClassificationSerum-free
FormLiquid
Product TypeCell Culture Supplement
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C) and protect from light.