4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine
Solubility
Reacts with water.
Notes
Moisture Sensitive. Protect from heat. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas. Incompatible with alcohols, amines and heat.
RUO – Research Use Only