Methyl 3-methoxyphenylacetate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-methoxyphenylacetate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Sparingly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Keith O. Hallinan; Toshio Honda. Enantiodivergent Synthesis of the Key Intermediate for Aphanorphine by Chemoenzymatic Process. Tetrahedron. 1995, 51 (45), 12211-12216.
- Timothy A Blizzard; Jerry D Morgan; Ralph T Mosley; Elizabeth T Birzin; Katalin Frisch; Susan P Rohrer; Milton L Hammond. 2-Phenylspiroindenes: a novel class of selective estrogen receptor modulators (SERMs). Bioorganic & Medicinal Chemistry Letters. 2003, 13 (3), 479-483.