2,3,6-Trifluorophenol, 98%
2,3,6-Trifluorophenol, 98%
2,3,6-Trifluorophenol, 98%
2,3,6-Trifluorophenol, 98%
Thermo Scientific Chemicals

2,3,6-Trifluorophenol, 98%

CAS: 113798-74-6 | C6H3F3O | 148.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20379.06
also known as B20379-06
Price (EUR)
117,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
117,00
Each
Add to cart
Chemical Identifiers
CAS113798-74-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream
Melting Point (clear melt)28.0-36.0°C
FormFused solid or lumpy powder
Identification (FTIR)Conforms
2,3,6-Trifluorophenol may be employed as model compound to investigate the site-selective functionalization of halogen-bearing phenols by organometallic approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trifluorophenol may be employed as model compound to investigate the site-selective functionalization of halogen-bearing phenols by organometallic approach.

Solubility
Soluble in water (47.267 mg/L est.)

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Marzi E and Schlosser M. The site-selective functionalization of halogen-bearing phenols: an exercise in diversity-oriented organometallic synthesis.Tetrahedron.2005,61(13), 3393-3401.
  2. M G Boersma, et al. 19F NMR metabolomics for the elucidation of microbial degradation pathways of fluorophenols.J Ind Microbiol Biotechnol.2001,26(1-2), 22-34.