N.N'-Bis(2-hydroxyethyl)oxamide, 99%
N.N'-Bis(2-hydroxyethyl)oxamide, 99%
N.N'-Bis(2-hydroxyethyl)oxamide, 99%
Thermo Scientific Chemicals

N.N'-Bis(2-hydroxyethyl)oxamide, 99%

CAS: 1871-89-2 | C6H12N2O4 | 176.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20458.22
also known as B20458-22
Price (EUR)
441,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
441,00
Each
Chemical Identifiers
CAS1871-89-2
IUPAC NameN,N'-bis(2-hydroxyethyl)ethanediamide
Molecular FormulaC6H12N2O4
InChI KeyFPQJEXTVQZHURJ-UHFFFAOYSA-N
SMILESOCCNC(=O)C(=O)NCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay (GC)>98.5%
N.N'-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N.N′-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S Kanda.; K Yamashita.; K Ohkawa. A proton conductive coordination polymer. I.[N, N'-bis (2-hydroxyethyl)-dithiooxamido] copper (II). Bulletin of the Chemical Society of Japan. 197952 ( 11), 3296-3301.
  2. I Van Poucke.; M Herman. Spectrophotometric determination of the dissociation constants of n. n'-bis (2-hydroxyethyl) dithio-oxamide. Analytica Chimica Acta. 196430 ( 1), 569-575.