Specifications
CAS72760-85-1
Chemical Name or Material3-Amino-5-methylthio-1H-pyrazole-4-carbonitrile
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight154.19
Health Hazard 1H302+H312+H332-H315-H319-H335
View more