N-Fmoc-L-leucine, 98%
N-Fmoc-L-leucine, 98%
N-Fmoc-L-leucine, 98%
Thermo Scientific Chemicals

N-Fmoc-L-leucine, 98%

CAS: 35661-60-0 | C21H23NO4 | 353.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21040.06
also known as B21040-06
Price (EUR)
39,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
39,60
Each
Chemical Identifiers
CAS35661-60-0
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid
Molecular FormulaC21H23NO4
InChI KeyCBPJQFCAFFNICX-TWYLJJHKNA-N
SMILESCC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (non-U.S. sourced material)
Melting Point (clear melt)150.0-157.0°C
Appearance (Color)White
Assay from Supplier's CofA≥97.5% (HPLC, U.S. sourced material)
Optical Rotation-25.0 ± 1.5° (C=1 in DMF)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only