Thermo Scientific Chemicals

Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers, tech., Thermo Scientific Chemicals

Catalog number: B21500.18
50 g, Each
Thermo Scientific Chemicals

Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers, tech., Thermo Scientific Chemicals

Catalog number: B21500.18
50 g, Each
Quantity
Catalog number: B21500.18
also known as B21500-18
Price (EUR)

Chemical Identifiers

CAS
374-76-5
IUPAC Name
1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris(trifluoromethyl)cyclohexane
Molecular Formula
C9F18
InChI Key
MGOFOLPXKULBGG-UHFFFAOYSA-N
SMILES
FC(F)(F)C1(F)C(F)(F)C(F)(C(F)(F)F)C(F)(F)C(F)(C(F)(F)F)C1(F)F
Appearance (Color)
Clear, colourless
Form
liquid
Assay (GC)
>98% (Sum of isomers)

Description

It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text