Thermo Scientific Chemicals

Bismuth(III) sulfate, 99%, Thermo Scientific Chemicals

Catalog number: B22007.14
25 g, Each
Thermo Scientific Chemicals

Bismuth(III) sulfate, 99%, Thermo Scientific Chemicals

Catalog number: B22007.14
25 g, Each
Quantity
Catalog number: B22007.14
also known as B22007-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7787-68-0
Elemental Analysis
Lead, Pb: ≤0.03% (≤300ppm)
Chloride content
<0.006% (<60ppm)
Elemental Analysis
Arsenic, As: ≤0.001% (≤10ppm)
Elemental Analysis
Copper, Cu: <0.006% (<60ppm)
Elemental Analysis
Iron, Fe: ≤0.002% (≤20ppm)

Description

The esterification of thujopsene with bismuth(III) sulfate in carboxylic acids yeilds esters in 30%. Bismuth(III) sulfateis used as analytics reagents and medicine, it is also used in pharmaceutical industry. It is used to determine other metal sulphate salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The esterification of thujopsene with bismuth(III) sulfate in carboxylic acids yeilds esters in 30%. Bismuth(III) sulfateis used as analytics reagents and medicine, it is also used in pharmaceutical industry. It is used to determine other metal sulphate salts.

Solubility
Soluble in acid, insoluble in water and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text