Thermo Scientific Chemicals

Methyl 3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: B22129.14
25 g, Each
Thermo Scientific Chemicals

Methyl 3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: B22129.14
25 g, Each
Quantity
Catalog number: B22129.14
also known as B22129-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
5118-06-9
IUPAC Name
(2Z)-2-[hydroxy(methoxy)methylidene]-2,3-dihydrothiophen-3-one
Molecular Formula
C6H6O3S
InChI Key
FYOPNADYSPZUAT-WAYWQWQTSA-N
SMILES
COC(O)=C1/SC=CC1=O
Assay (GC)
≥96.0%
Appearance (Color)
White to pale yellow or pale cream to pale brown
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
41.0-47.0?C

Description

Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention