Aminoacetonitrile sulfate, 97%
Aminoacetonitrile sulfate, 97%
Aminoacetonitrile sulfate, 97%
Aminoacetonitrile sulfate, 97%
Thermo Scientific Chemicals

Aminoacetonitrile sulfate, 97%

CAS: 5466-22-8 | C4H8N8·H2SO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B22155.36
also known as B22155-36
Price (EUR)
331,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
331,00
Each
Add to cart
Chemical Identifiers
CAS5466-22-8
IUPAC Namebis(cyanomethanaminium) sulfate
Molecular FormulaC4H10N4O4S
InChI KeyNQHWLBZZBSYIPI-UHFFFAOYSA-N
SMILES[NH3+]CC#N.[NH3+]CC#N.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale brown
FormPowder or chunks
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only
Hidesaburo Hanafusa; Shiro Akabori. Polymerization of Aminoacetonitrile. Bulletin of the Chemical Society of Japan. 1959, 32 (6), 626-630.