4-(Trifluoromethyl)benzylamine, 97%
4-(Trifluoromethyl)benzylamine, 97%
4-(Trifluoromethyl)benzylamine, 97%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzylamine, 97%

CAS: 3300-51-4 | C8H8F3N | 175.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22745.03
also known as B22745-03
Price (EUR)
42,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
42,10
Each
Chemical Identifiers
CAS3300-51-4
IUPAC Name1-[4-(trifluoromethyl)phenyl]methanamine
Molecular FormulaC8H8F3N
InChI KeyPRDBLLIPPDOICK-UHFFFAOYSA-N
SMILESNCC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only