Dichloromethylvinylsilane, 97%
Dichloromethylvinylsilane, 97%
Dichloromethylvinylsilane, 97%
Dichloromethylvinylsilane, 97%
Thermo Scientific Chemicals

Dichloromethylvinylsilane, 97%

CAS: 124-70-9 | C3H6Cl2Si | 141.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B23119.36
also known as B23119-36
Price (EUR)
120,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
120,00
Each
Add to cart
Chemical Identifiers
CAS124-70-9
IUPAC Namedichloro(ethenyl)methylsilane
Molecular FormulaC3H6Cl2Si
InChI KeyYLJJAVFOBDSYAN-UHFFFAOYSA-N
SMILESC[Si](Cl)(Cl)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow to brown
Refractive Index1.4270-1.4320 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only