4-Methoxybenzophenone and its analogues showed diverse plant growth-regulating actions such as inhibition of shoot and root growth, induction of chlorosis, and a disturbance in phototropism or geotropism. It is also used as a photo polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzophenone and its analogues showed diverse plant growth-regulating actions such as inhibition of shoot and root growth, induction of chlorosis, and a disturbance in phototropism or geotropism. It is also used as a photo polymerization catalyst.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
RUO – Research Use Only
General References:
- Francisco Boscá; Gonzalo Cosa; Miguel A. Miranda and J. C. Scaiano. Inversion of 4-methoxybenzophenone triplet in aqueous solutions. Photochemical & Photobiological Sciences. 2002, 1 (9), 704-708.
- T. Suthan; N.P. Rajesh; C.K. Mahadevan; G. Bhagavannarayana. Growth and characterization of 2-hydroxy-4-methoxybenzophenone single crystal using modified vertical Bridgman technique. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2011, 78 (2), 771-776.