3-(4-Morpholinyl)propylamine, 98%
3-(4-Morpholinyl)propylamine, 98%
3-(4-Morpholinyl)propylamine, 98%
Thermo Scientific Chemicals

3-(4-Morpholinyl)propylamine, 98%

CAS: 123-00-2 | C7H16N2O | 144.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number B23485.30
also known as B23485-30
Price (EUR)
251,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
251,00
Each
Chemical Identifiers
CAS123-00-2
IUPAC Name3-(morpholin-4-yl)propan-1-amine
Molecular FormulaC7H16N2O
InChI KeyUIKUBYKUYUSRSM-UHFFFAOYSA-N
SMILESNCCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5% (U.K. specification)
CommentPurchased in U.K. and U.S.
Assay (Non-aqueous acid-base Titration)≥97.5% to ≤102.5% (U.K. specification)
View more
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.
RUO – Research Use Only