4-Amino-1-naphthol hydrochloride, tech. 90%
4-Amino-1-naphthol hydrochloride, tech. 90%
4-Amino-1-naphthol hydrochloride, tech. 90%
4-Amino-1-naphthol hydrochloride, tech. 90%
Thermo Scientific Chemicals

4-Amino-1-naphthol hydrochloride, tech. 90%

CAS: 5959-56-8 | C10H10ClNO | 195.646 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23978.14
also known as B23978-14
Price (EUR)
229,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
229,00
Each
Add to cart
Chemical Identifiers
CAS5959-56-8
IUPAC Name4-hydroxynaphthalen-1-aminium chloride
Molecular FormulaC10H10ClNO
InChI KeyFDBQTRARWCKEJY-UHFFFAOYSA-N
SMILES[Cl-].[NH3+]C1=C2C=CC=CC2=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)≥88.0%
Assay (Titration ex Chloride)≥88.0%
Appearance (Color)Grey or brown to purple
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only