N-Methylisatoic anhydride, tech. 90%
N-Methylisatoic anhydride, tech. 90%
N-Methylisatoic anhydride, tech. 90%
Thermo Scientific Chemicals

N-Methylisatoic anhydride, tech. 90%

CAS: 10328-92-4 | C9H7NO3 | 177.159 g/mol
Have Questions?
Catalog NumberQuantity
B24099.22
also known as B24099-22
100 g
Catalog number B24099.22
also known as B24099-22
Price (EUR)
80,40
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
80,40
Each
Chemical Identifiers
CAS10328-92-4
IUPAC Name1-methyl-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Molecular FormulaC9H7NO3
InChI KeyKJMRWDHBVCNLTQ-UHFFFAOYSA-N
SMILESCN1C(=O)OC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Assay (HPLC)≥88.0%
Appearance (Color)White to brown or pale grey
Assay (Aqueous acid-base Titration)≥88.0 to ≤112.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only