4-Bromo-2-fluorobenzenesulfonyl chloride, 98%
4-Bromo-2-fluorobenzenesulfonyl chloride, 98%
4-Bromo-2-fluorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzenesulfonyl chloride, 98%

CAS: 216159-03-4 | C6H3BrClFO2S | 273.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24352.14
also known as B24352-14
Price (EUR)
177,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
177,00
Each
Chemical Identifiers
CAS216159-03-4
IUPAC Name4-bromo-2-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H3BrClFO2S
InChI KeyXNYBZLRIUHNRQY-UHFFFAOYSA-N
SMILESFC1=C(C=CC(Br)=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)62.5-68.5?C
Appearance (Color)White to pale cream to pale yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only