Thermo Scientific Chemicals

3-Methyl-2-butanone, 98%, Thermo Scientific Chemicals

Catalog number: B24527.AK
250 mL, Each
Thermo Scientific Chemicals

3-Methyl-2-butanone, 98%, Thermo Scientific Chemicals

Catalog number: B24527.AK
250 mL, Each
Quantity
Catalog number: B24527.AK
also known as B24527-AK
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
563-80-4
IUPAC Name
3-methylbutan-2-one
Molecular Formula
C5H10O
InChI Key
SYBYTAAJFKOIEJ-UHFFFAOYSA-N
SMILES
CC(C)C(C)=O
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.3865-1.3905 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.

Solubility
Soluble in water (8.21 mg/ml at 20°C), ether, alcohol, acetone, and ethanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 2
    Label Elements
    Signal Word

    Danger

    Physical Hazards

    H225 - Highly flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P240 - Ground and bond container and receiving equipment