3-Bromo-4-methylaniline, 97%
3-Bromo-4-methylaniline, 97%
3-Bromo-4-methylaniline, 97%
3-Bromo-4-methylaniline, 97%
Thermo Scientific Chemicals

3-Bromo-4-methylaniline, 97%

CAS: 7745-91-7 | C7H8BrN | 186.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24670.06
also known as B24670-06
Price (EUR)
47,60
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
47,60
Each
Add to cart
Chemical Identifiers
CAS7745-91-7
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6100-1.6150 @ 20?C
Assay (GC)≥96.0%
Appearance (Color)Clear yellow to red to brown to dark brown
CommentMay darken on storage
FormLiquid

3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger?s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger′s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Matthew B Prendergast.; Phillip A Cooper.; Benjamin B Kirk.; Gabriel da Silva.; Stephen J Blanksby.; Adam J Trevitt. Hydroxyl radical formation in the gas phase oxidation of distonic 2-methylphenyl radical cations. Physical Chemistry Chemical Physics. 2013, 15 (47), 20577-20584.
  2. M. S. Carpenter.; W. M. Easter. The Isopropyl Cresols. J. Org. Chem. 1955, 20 (4), 401-411.