4-Methylphenylacetone, 97+%
4-Methylphenylacetone, 97+%
4-Methylphenylacetone, 97+%
4-Methylphenylacetone, 97+%
Thermo Scientific Chemicals

4-Methylphenylacetone, 97+%

CAS: 2096-86-8 | C10H12O | 148.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24728.03
also known as B24728-03
Price (EUR)
30,40
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
30,40
Each
Add to cart
Chemical Identifiers
CAS2096-86-8
IUPAC Name1-(4-methylphenyl)propan-2-one
Molecular FormulaC10H12O
InChI KeyNOXKUHSBIXPZBJ-UHFFFAOYSA-N
SMILESCC(=O)CC1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
FormLiquid
Assay (GC)≥97.0%
Refractive Index1.5090-1.5150 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only