Ethyl cyclohexylideneacetate, 98%
Ethyl cyclohexylideneacetate, 98%
Ethyl cyclohexylideneacetate, 98%
Thermo Scientific Chemicals

Ethyl cyclohexylideneacetate, 98%

CAS: 1552-92-7 | C10H16O2 | 168.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25073.14
also known as B25073-14
Price (EUR)
211,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
211,00
Each
Chemical Identifiers
CAS1552-92-7
IUPAC Nameethyl 2-cyclohexylideneacetate
Molecular FormulaC10H16O2
InChI KeyMCWDXHYYYNGYGK-UHFFFAOYSA-N
SMILESCCOC(=O)C=C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4770-1.4810 @ 20?C
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl acetate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only