2,4-Dimethyl-6-nitrophenol, 98%
2,4-Dimethyl-6-nitrophenol, 98%
2,4-Dimethyl-6-nitrophenol, 98%
Thermo Scientific Chemicals

2,4-Dimethyl-6-nitrophenol, 98%

CAS: 14452-34-7 | C8H9NO3 | 167.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25205.14
also known as B25205-14
Price (EUR)
163,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
163,00
Each
Chemical Identifiers
CAS14452-34-7
IUPAC Name2,4-dimethyl-6-nitrophenol
Molecular FormulaC8H9NO3
InChI KeyKJRCHILWKQLEBC-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(O)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)66.0-72.0?C
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only