4-(Trifluoromethyl)benzenesulfonamide, 97%
4-(Trifluoromethyl)benzenesulfonamide, 97%
4-(Trifluoromethyl)benzenesulfonamide, 97%
4-(Trifluoromethyl)benzenesulfonamide, 97%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzenesulfonamide, 97%

CAS: 830-43-3 | C7H6F3NO2S | 225.185 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number B25243.03
also known as B25243-03
Price (EUR)
33,80
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
33,80
Each
Add to cart
Chemical Identifiers
CAS830-43-3
IUPAC Name4-(trifluoromethyl)benzene-1-sulfonamide
Molecular FormulaC7H6F3NO2S
InChI KeyTVHXQQJDMHKGGK-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)174.5-183.5?C
Assay (GC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only