2,5-Bis(trifluoromethyl)benzoic acid, 98%
2,5-Bis(trifluoromethyl)benzoic acid, 98%
2,5-Bis(trifluoromethyl)benzoic acid, 98%
Thermo Scientific Chemicals

2,5-Bis(trifluoromethyl)benzoic acid, 98%

CAS: 42580-42-7 | C9H4F6O2 | 258.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26184.06
also known as H26184-06
Price (EUR)
63,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
63,00
Each
Chemical Identifiers
CAS42580-42-7
IUPAC Name2,5-bis(trifluoromethyl)benzoic acid
Molecular FormulaC9H4F6O2
InChI KeyPINBPLCVZSKLTF-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC=C1C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)75.0-81.0?C
Appearance (Color)Pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only