4-n-Octylphenol, 99%
4-n-Octylphenol, 99%
4-n-Octylphenol, 99%
Thermo Scientific Chemicals

4-n-Octylphenol, 99%

CAS: 1806-26-4 | C14H22O | 206.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26396.06
also known as H26396-06
Price (EUR)
93,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
93,40
Each
Chemical Identifiers
CAS1806-26-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Shigeru Kohtani.; Masaya Koshiko.; Akihiko Kudo.; Kunihiro Tokumura.; Yasuhito Ishigaki.; Akira Toriba.; Kazuichi Hayakawa.; Ryoichi Nakagaki. Photodegradation of 4-alkylphenols using BiVO4 photocatalyst under irradiation with visible light from a solar simulator. Applied Catalysis B: Environmental. 2003, 46 (3), 573-586.
  2. T. Tanizaki.; K. Kadokami.; R. Shinohara. Bulletin of Environmental Contamination and Toxicology. Bulletin of Environmental Contamination and Toxicology. 2002, 68 (5), 732-739.