2-Hydroxypyrimidine-5-boronic acid pinacol ester, 95%
2-Hydroxypyrimidine-5-boronic acid pinacol ester, 95%
2-Hydroxypyrimidine-5-boronic acid pinacol ester, 95%
Thermo Scientific Chemicals

2-Hydroxypyrimidine-5-boronic acid pinacol ester, 95%

CAS: 1073354-84-3 | C10H15BN2O3 | 222.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26976.03
also known as H26976-03
Price (EUR)
722,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
722,00
Each
Chemical Identifiers
CAS1073354-84-3
IUPAC Name5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-dihydropyrimidin-2-one
Molecular FormulaC10H15BN2O3
InChI KeyJRCSRFGCQSLWNZ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CNC(=O)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥94.0%
Appearance (Color)White to pale yellow or pale cream
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only