5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%
Thermo Scientific Chemicals

5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%

CAS: 51516-68-8 | C10H7ClN4 | 218.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27287.06
also known as H27287-06
Price (EUR)
115,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
115,00
Each
Chemical Identifiers
CAS51516-68-8
IUPAC Name5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile
Molecular FormulaC10H7ClN4
InChI KeyVRKRSWGTAMOYEV-UHFFFAOYSA-N
SMILESNC1=C(C=NN1C1=CC=CC(Cl)=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
Melting Point (clear melt)181.0-190.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only