3-Bromo-4-fluorobenzyl alcohol, 97%
3-Bromo-4-fluorobenzyl alcohol, 97%
3-Bromo-4-fluorobenzyl alcohol, 97%
3-Bromo-4-fluorobenzyl alcohol, 97%
Thermo Scientific Chemicals

3-Bromo-4-fluorobenzyl alcohol, 97%

CAS: 77771-03-0 | C7H6BrFO | 205.026 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H30604.06
also known as H30604-06
Price (EUR)
30,20
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
30,20
Each
Add to cart
Chemical Identifiers
CAS77771-03-0
IUPAC Name(3-bromo-4-fluorophenyl)methanaminium
Molecular FormulaC7H8BrFN
InChI KeyWIRTWOIWFFCEPB-UHFFFAOYSA-O
SMILES[NH3+]CC1=CC=C(F)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.5590-1.5640 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only