N-Boc-N-methyl-L-alanine, 98%
N-Boc-N-methyl-L-alanine, 98%
N-Boc-N-methyl-L-alanine, 98%
N-Boc-N-methyl-L-alanine, 98%
Thermo Scientific™

N-Boc-N-methyl-L-alanine, 98%

CAS: 16948-16-6 | C9H17NO4 | 203.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31317.03
also known as H31317-03
Price (EUR)
53,72
Online Exclusive
63,20
Save 9,48 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
53,72
Online Exclusive
63,20
Save 9,48 (15%)
Each
Add to cart
Chemical Identifiers
CAS16948-16-6
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
Molecular FormulaC9H17NO4
InChI KeyVLHQXRIIQSTJCQ-LURJTMIESA-N
SMILESC[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Optical Rotation-30.4 ± 0.3? (C=1 in ethanol)
Melting Point (clear melt)88.0-94.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only