Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H32065.03
also known as H32065-03
1 g
Catalog number H32065.03
also known as H32065-03
Price (EUR)
134,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
134,00
Each
Chemical Identifiers
CAS306960-67-8
IUPAC Nameethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
Molecular FormulaC9H9F3N2O2
InChI KeyLYEKBQSEKSEXLT-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CN=C(N=C1C)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear pale yellow
Assay (GC)≥96.0%
Refractive Index1.4415-1.4465 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only