1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine, 96%
1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine, 96%
1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine, 96%
Thermo Scientific Chemicals

1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine, 96%

CAS: 84162-82-3 | C14H15F2NO2 | 267.276 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H33058.03
also known as H33058-03
Price (EUR)
80,80
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
80,80
Each
Chemical Identifiers
CAS84162-82-3
IUPAC Name1-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethan-1-one
Molecular FormulaC14H15F2NO2
InChI KeyUGPWQEIGJWVJDR-UHFFFAOYSA-N
SMILESCC(=O)N1CCC(CC1)C(=O)C1=C(F)C=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)94.0-103.0?C
Appearance (Color)Pale cream
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only