Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 96%
Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 96%
Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 96%
Thermo Scientific Chemicals

Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 96%

CAS: 4815-29-6 | C10H13NO2S | 211.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H33196.14
also known as H33196-14
Price (EUR)
267,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
267,00
Each
Chemical Identifiers
CAS4815-29-6
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)88.0-97.0?C
Appearance (Color)Pale yellow to brown
FormPowder
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only