2-Bromo-4-chloropyridine, 97%
2-Bromo-4-chloropyridine, 97%
2-Bromo-4-chloropyridine, 97%
Thermo Scientific Chemicals

2-Bromo-4-chloropyridine, 97%

CAS: 22918-01-0 | C5H3BrClN | 192.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33698.03
also known as H33698-03
Price (EUR)
37,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
37,40
Each
Chemical Identifiers
CAS22918-01-0
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.5855-1.5905 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only