Specifications
CAS125903-81-3
Chemical Name or Material3-(Difluoromethoxy)benzyl alcohol
Formula Weight174.15
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more