3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H51709.06
also known as H51709-06
5 g
Catalog number H51709.06
also known as H51709-06
Price (EUR)
191,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
191,00
Each
Add to cart
Specifications
CAS1257651-17-4
Chemical Name or Material3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester
Formula Weight376.26
IUPAC Nametert-butyl N-[2-oxo-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anilino]ethyl]carbamate
InChI KeyIYTPRTWBNLJZLK-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only